Assay Details
| Gene Symbol: FEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122627.5 | XP_003122675.1 | 654 | 61 | ||
| GenBank mRNA | AK236480.1 | - | 1 | 320 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: FEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122627.5 | XP_003122675.1 | 654 | 61 | ||
| GenBank mRNA | AK236480.1 | - | 1 | 320 | 61 | |
Set Membership: |
|