Assay Details
Gene Symbol: FEZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003125247.6 | XP_003125295.4 | 397 | 89 | ||
XM_003481247.4 | XP_003481295.2 | 403 | 89 | |||
XM_021087642.1 | XP_020943301.1 | 397 | 89 | |||
XM_021087643.1 | XP_020943302.1 | 397 | 89 | |||
GenBank mRNA | AK236524.1 | - | 2 - 3 | 372 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
