Assay Details
Gene Symbol: LOC100523244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126024.6 | XP_003126072.1 | 454 | 69 | ||
XM_005655504.3 | XP_005655561.1 | 455 | 69 | |||
XM_005655505.3 | XP_005655562.1 | 455 | 69 | |||
XM_005655506.3 | XP_005655563.1 | 455 | 69 | |||
XM_005655507.3 | XP_005655564.1 | 454 | 69 | |||
XM_005655508.3 | XP_005655565.1 | 454 | 69 | |||
XM_005655509.3 | XP_005655566.1 | 451 | 69 | |||
XM_005655510.3 | XP_005655567.1 | 455 | 69 | |||
XM_013988202.2 | XP_013843656.1 | 454 | 69 | |||
XM_021091492.1 | XP_020947151.1 | 285 | 69 | |||
GenBank mRNA | AK236660.1 | - | 3 - 4 | 352 | 69 | |
AK391789.1 | - | 3 - 4 | 431 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
