Assay Details
Gene Symbol: LOC100523244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126024.6 | XP_003126072.1 | 1349 | 76 | ||
XM_005655504.3 | XP_005655561.1 | 1411 | 76 | |||
XM_005655505.3 | XP_005655562.1 | 1411 | 76 | |||
XM_005655506.3 | XP_005655563.1 | 1411 | 76 | |||
XM_005655507.3 | XP_005655564.1 | 1349 | 76 | |||
XM_005655508.3 | XP_005655565.1 | 1349 | 76 | |||
XM_005655509.3 | XP_005655566.1 | 1346 | 76 | |||
XM_021091492.1 | XP_020947151.1 | 1180 | 76 | |||
GenBank mRNA | AK236660.1 | - | 8 - 9 | 1247 | 76 | |
AK391789.1 | - | 8 - 9 | 1326 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
