Assay Details
Gene Symbol: REM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005654242.3 | XP_005654299.3 | 495 | 77 | ||
XM_013983226.2 | XP_013838680.2 | 669 | 77 | |||
XM_013983232.2 | XP_013838686.2 | 495 | 77 | |||
GenBank mRNA | AK236667.1 | - | 2 - 3 | 618 | 77 | |
AK351161.1 | - | 2 - 3 | 601 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
