Assay Details
Gene Symbol: LGALS3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021066516.1 | XP_020922175.1 | 550 | 65 | ||
GenBank mRNA | AK236708.1 | - | 4 - 5 | 530 | 65 | |
AK392852.1 | - | 4 - 5 | 499 | 65 | ||
AK397864.1 | - | 4 - 5 | 474 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
