Assay Details
| Gene Symbol: PYCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090543.1 | XP_020946202.1 | 266 | 74 | ||
| GenBank mRNA | AK236710.1 | - | 2 - 3 | 191 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PYCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090543.1 | XP_020946202.1 | 266 | 74 | ||
| GenBank mRNA | AK236710.1 | - | 2 - 3 | 191 | 74 | |
Set Membership: |
|