Assay Details
Gene Symbol: MAGOHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013988371.2 | XP_013843825.1 | 285 | 96 | ||
GenBank mRNA | AK236808.1 | - | 2 - 3 | 483 | 96 | |
AY609501.1 | - | 1 - 2 | 107 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
