Assay Details
Gene Symbol: EGLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021073580.1 | XP_020929239.1 | 1525 | 94 | ||
GenBank mRNA | AK236874.1 | - | 1 - 2 | 43 | 94 | |
AK393674.1 | - | 4 - 5 | 483 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
