Assay Details
| Gene Symbol: SAP18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065443.1 | XP_020921102.1 | 4510 | 120 | ||
| GenBank mRNA | AK236951.1 | - | 2 - 3 | 206 | 120 | |
| AK346604.1 | - | 2 - 3 | 202 | 120 | ||
| AK351199.1 | - | 2 - 3 | 208 | 120 | ||
| AK351228.1 | - | 2 - 3 | 689 | 120 | ||
| AK399832.1 | - | 2 - 3 | 251 | 120 | ||
More Information
Set Membership: |
|

SAP18
Genomic Map