Assay Details
Gene Symbol: SLC25A17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126004.4 | XP_003126052.1 | 600 | 70 | ||
GenBank mRNA | AK237005.1 | - | 3 - 4 | 431 | 70 | |
AK237571.1 | - | 5 - 6 | 605 | 70 | ||
AK239016.1 | - | 5 - 6 | 531 | 70 | ||
AK389811.1 | - | 5 - 6 | 573 | 70 | ||
AY609429.1 | - | 5 - 6 | 536 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
