Assay Details
| Gene Symbol: NMNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095304.1 | XP_020950963.1 | 769 | 79 | ||
| XM_021095305.1 | XP_020950964.1 | 820 | 79 | |||
| XM_021095306.1 | XP_020950965.1 | 1251 | 79 | |||
| XM_021095307.1 | XP_020950966.1 | 819 | 79 | |||
| GenBank mRNA | AK237038.1 | - | 4 - 5 | 517 | 79 | |
More Information
Set Membership: |
|

NMNAT1
Genomic Map