Assay Details
Gene Symbol: FES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001929355.4 | XP_001929390.1 | 2506 | 64 | ||
XM_021098925.1 | XP_020954584.1 | 1903 | 64 | |||
GenBank mRNA | AK237219.1 | - | 13 - 14 | 1756 | 64 | |
AK394391.1 | - | 13 - 14 | 1790 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
