Assay Details
| Gene Symbol: CXCL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129101.3 | XP_003129149.1 | 281 | 70 | ||
| GenBank mRNA | AK237249.1 | - | 2 - 3 | 249 | 70 | |
| AK347056.1 | - | 2 - 3 | 244 | 70 | ||
More Information
Set Membership: |
|

CXCL13
Genomic Map