Assay Details
| Gene Symbol: PXMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077363.1 | XP_020933022.1 | 312 | 99 | ||
| XM_021077364.1 | XP_020933023.1 | 235 | 99 | |||
| GenBank mRNA | AK237268.1 | - | 1 - 2 | 227 | 99 | |
| AK347658.1 | - | 1 - 2 | 178 | 99 | ||
| AK348336.1 | - | 1 - 2 | 171 | 99 | ||
More Information
Set Membership: |
|

PXMP4
Genomic Map