Assay Details
| Gene Symbol: NIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005663170.3 | XP_005663227.1 | 452 | 67 | ||
| XM_005663172.3 | XP_005663229.1 | 589 | 67 | |||
| XM_005663174.3 | XP_005663231.1 | 693 | 67 | |||
| XM_005663175.3 | XP_005663232.1 | 744 | 67 | |||
| GenBank mRNA | AK237318.1 | - | 3 - 4 | 439 | 67 | |
| AK237892.1 | - | 3 - 4 | 476 | 67 | ||
| AK392012.1 | - | 3 - 4 | 462 | 67 | ||
| AK399212.1 | - | 3 - 4 | 411 | 67 | ||
More Information
Set Membership: |
|

NIT1
Genomic Map