Assay Details
| Gene Symbol: YIPF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087657.1 | XP_020943316.1 | 338 | 91 | ||
| GenBank mRNA | AK237364.1 | - | 1 - 2 | 295 | 91 | |
| AK390447.1 | - | 1 - 2 | 328 | 91 | ||
| AY609471.1 | - | 1 - 2 | 255 | 91 | ||
More Information
Set Membership: |
|

YIPF4
Genomic Map