Assay Details
| Gene Symbol: PKIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013986820.2 | XP_013842274.1 | 752 | 92 | ||
| XM_021088778.1 | XP_020944437.1 | 735 | 92 | |||
| XM_021088787.1 | XP_020944446.1 | 593 | 92 | |||
| XM_021088793.1 | XP_020944452.1 | 746 | 92 | |||
| XM_021088801.1 | XP_020944460.1 | 628 | 92 | |||
| XM_021088811.1 | XP_020944470.1 | 602 | 92 | |||
| XM_021088819.1 | XP_020944478.1 | 409 | 92 | |||
| GenBank mRNA | AK237739.1 | - | 4 - 5 | 482 | 92 | |
| AK237740.1 | - | 4 - 5 | 482 | 92 | ||
More Information
Set Membership: |
|

PKIB
Genomic Map