Assay Details
| Gene Symbol: GPRASP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135261.4 | XP_003135309.1 | 3495 | 101 | ||
| XM_005657879.3 | XP_005657936.1 | 5471 | 101 | |||
| XM_005657880.3 | XP_005657937.1 | 3580 | 101 | |||
| XM_005657881.3 | XP_005657938.1 | 3659 | 101 | |||
| XM_013990969.2 | XP_013846423.1 | 3699 | 101 | |||
| XM_013990970.2 | XP_013846424.1 | 3499 | 101 | |||
| XM_013990971.2 | XP_013846425.1 | 3503 | 101 | |||
| XM_021079933.1 | XP_020935592.1 | 3589 | 101 | |||
| XM_021079934.1 | XP_020935593.1 | 3543 | 101 | |||
| XM_021079935.1 | XP_020935594.1 | 3418 | 101 | |||
| GenBank mRNA | AK237771.1 | - | 1 | 1316 | 101 | |
| AK349134.1 | - | 4 | 3371 | 101 | ||
More Information
Set Membership: |
|

GPRASP2
Genomic Map