Assay Details
Gene Symbol: LY6E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021088765.1 | XP_020944424.1 | 441 | 63 | ||
XM_021088766.1 | XP_020944425.1 | 306 | 63 | |||
XM_021088767.1 | XP_020944426.1 | 476 | 63 | |||
XM_021088768.1 | XP_020944427.1 | 1054 | 63 | |||
XM_021088769.1 | XP_020944428.1 | 456 | 63 | |||
GenBank mRNA | AK237832.1 | - | 3 - 4 | 450 | 63 | |
AK350824.1 | - | 3 - 4 | 301 | 63 | ||
KJ820673.1 | - | 2 - 3 | 178 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
