Assay Details
| Gene Symbol: IRAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003480735.4 | XP_003480783.2 | 5226 | 91 | ||
| XM_013994558.2 | XP_013850012.1 | 5265 | 91 | |||
| XM_013994559.2 | XP_013850013.1 | 5152 | 91 | |||
| XM_013994560.2 | XP_013850014.1 | 5111 | 91 | |||
| XM_013994562.2 | XP_013850016.1 | 4452 | 91 | |||
| XM_013994564.2 | XP_013850018.1 | 5222 | 91 | |||
| XM_013994565.2 | XP_013850019.1 | 5016 | 91 | |||
| XM_021083326.1 | XP_020938985.1 | 4370 | 91 | |||
| XM_021083328.1 | XP_020938987.1 | 4533 | 91 | |||
| XM_021083329.1 | XP_020938988.1 | 4475 | 91 | |||
| XM_021083330.1 | XP_020938989.1 | 4345 | 91 | |||
| XM_021083332.1 | XP_020938991.1 | 5056 | 91 | |||
| XM_021083333.1 | XP_020938992.1 | 4983 | 91 | |||
| XM_021083334.1 | XP_020938993.1 | 4955 | 91 | |||
| GenBank mRNA | AK238022.1 | - | 1 | 1318 | 91 | |
More Information
Set Membership: |
|

IRAG1
Genomic Map