Assay Details
| Gene Symbol: ZFP36L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125190.6 | XP_003125238.1 | 1402 | 64 | ||
| GenBank mRNA | AK238088.1 | - | 1 | 718 | 64 | |
| AK348798.1 | - | 1 | 1928 | 64 | ||
More Information
Set Membership: |
|

ZFP36L2
Genomic Map