Assay Details
| Gene Symbol: MXRA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066565.1 | XP_020922224.1 | 899 | 86 | ||
| XM_021066568.1 | XP_020922227.1 | 893 | 86 | |||
| XM_021066569.1 | XP_020922228.1 | 812 | 86 | |||
| GenBank mRNA | AK238220.1 | - | 1 | 217 | 86 | |
| AK389423.1 | - | 1 | 273 | 86 | ||
| AK397747.1 | - | 1 | 306 | 86 | ||
More Information
Set Membership: |
|

MXRA7
Genomic Map