Assay Details
| Gene Symbol: COX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929331.5 | XP_001929366.1 | 973 | 57 | ||
| XM_021072321.1 | XP_020927980.1 | 722 | 57 | |||
| GenBank mRNA | AK231019.1 | - | 6 - 7 | 931 | 57 | |
More Information
Set Membership: |
|

COX15
Genomic Map