Assay Details
| Gene Symbol: CD164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924626.5 | XP_001924661.2 | 2207 | 88 | ||
| XM_021090988.1 | XP_020946647.1 | 2052 | 88 | |||
| GenBank mRNA | AK231031.1 | - | 1 | 1120 | 88 | |
More Information
Set Membership: |
|

CD164
Genomic Map