Assay Details
Gene Symbol: AURKAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021097450.1 | XP_020953109.1 | 280 | 68 | ||
XM_021097451.1 | XP_020953110.1 | 393 | 68 | |||
XM_021097452.1 | XP_020953111.1 | 322 | 68 | |||
XM_021097453.1 | XP_020953112.1 | 286 | 68 | |||
XM_021097454.1 | XP_020953113.1 | 300 | 68 | |||
GenBank mRNA | AK231191.1 | - | 1 - 2 | 456 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
