Assay Details
| Gene Symbol: FXYD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013998301.2 | XP_013853755.1 | 377 | 68 | ||
| XM_013998302.2 | XP_013853756.1 | 333 | 68 | |||
| XM_013998303.2 | XP_013853757.1 | 269 | 68 | |||
| XM_013998304.2 | XP_013853758.1 | 185 | 68 | |||
| XM_013998306.2 | XP_013853760.1 | 377 | 68 | |||
| XM_013998307.2 | XP_013853761.1 | 333 | 68 | |||
| XM_013998308.2 | XP_013853762.1 | 269 | 68 | |||
| XM_021097213.1 | XP_020952872.1 | 188 | 68 | |||
| XM_021097214.1 | XP_020952873.1 | 188 | 68 | |||
| XM_021097215.1 | XP_020952874.1 | 188 | 68 | |||
| GenBank mRNA | AK231364.1 | - | 2 - 3 | 712 | 68 | |
| AK352494.1 | - | 3 - 4 | 306 | 68 | ||
More Information
Set Membership: |
|

FXYD5
Genomic Map