Assay Details
| Gene Symbol: PGM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121933.6 | XP_003121981.4 | 2523 | 109 | ||
| GenBank mRNA | AK231404.1 | - | 1 | 122 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PGM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121933.6 | XP_003121981.4 | 2523 | 109 | ||
| GenBank mRNA | AK231404.1 | - | 1 | 122 | 109 | |
Set Membership: |
|