Assay Details
Gene Symbol: C2H11orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003480654.4 | XP_003480702.1 | 176 | 86 | ||
XM_013994015.2 | XP_013849469.1 | 164 | 86 | |||
XM_021082724.1 | XP_020938383.1 | 164 | 86 | |||
GenBank mRNA | AK231440.1 | - | 1 - 2 | 114 | 86 | |
AK390051.1 | - | 1 - 2 | 160 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003480654.4, XM_013994015.2, XM_021082724.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
