Assay Details
Gene Symbol: CBLIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003122682.3 | XP_003122730.2 | 609 | 67 | ||
GenBank mRNA | AK231492.1 | - | 4 - 5 | 568 | 67 | |
AK400404.1 | - | 4 - 5 | 568 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
