Assay Details
Gene Symbol: NSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003123703.4 | XP_003123751.1 | 531 | 91 | ||
GenBank mRNA | AK231501.1 | - | 3 - 4 | 455 | 91 | |
AK231798.1 | - | 3 - 4 | 465 | 91 | ||
AK397476.1 | - | 713 | 91 | |||
AK399125.1 | - | 3 - 4 | 470 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
