Assay Details
Gene Symbol: MAPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021088019.1 | XP_020943678.1 | 383 | 65 | ||
GenBank mRNA | AK231529.1 | - | 1 - 2 | 218 | 65 | |
AK391915.1 | - | 2 - 3 | 241 | 65 | ||
HM745137.1 | - | 1 - 2 | 144 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
