Assay Details
| Gene Symbol: YWHAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005672961.3 | XP_005673018.1 | 813 | 73 | ||
| GenBank mRNA | AK231552.1 | - | 4 - 5 | 790 | 73 | |
| AK231708.1 | - | 4 - 5 | 790 | 73 | ||
| AK348020.1 | - | 1 - 2 | 51 | 73 | ||
| AK392687.1 | - | 4 - 5 | 762 | 73 | ||
| AK397398.1 | - | 1 - 2 | 107 | 73 | ||
| AK397784.1 | - | 4 - 5 | 737 | 73 | ||
| AK398867.1 | - | 4 - 5 | 795 | 73 | ||
| AK399290.1 | - | 4 - 5 | 790 | 73 | ||
More Information
Set Membership: |
|

YWHAB
Genomic Map