Assay Details
Gene Symbol: DUSP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003483054.4 | XP_003483102.2 | 127 | 70 | ||
GenBank mRNA | AK238568.1 | - | 1 - 2 | 65 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003483054.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
