Assay Details
| Gene Symbol: PAN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005668294.3 | XP_005668351.1 | 5198 | 100 | ||
| XM_005668295.3 | XP_005668352.1 | 5195 | 100 | |||
| XM_005668297.3 | XP_005668354.1 | 5036 | 100 | |||
| XM_005668298.3 | XP_005668355.1 | 4994 | 100 | |||
| XM_013980413.2 | XP_013835867.1 | 4724 | 100 | |||
| XM_021065522.1 | XP_020921181.1 | 5073 | 100 | |||
| GenBank mRNA | AK238669.1 | - | 1 | 915 | 100 | |
More Information
Set Membership: |
|

PAN3
Genomic Map