Assay Details
| Gene Symbol: MFAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928118.5 | XP_001928153.1 | 816 | 62 | ||
| GenBank mRNA | AK238787.1 | - | 4 - 5 | 765 | 62 | |
| AK397307.1 | - | 4 - 5 | 789 | 62 | ||
More Information
Set Membership: |
|

MFAP1
Genomic Map