Assay Details
Gene Symbol: PIMREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003131878.4 | XP_003131926.2 | 786 | 70 | ||
GenBank mRNA | AK239230.1 | - | 4 - 5 | 726 | 70 | |
AK347529.1 | - | 4 - 5 | 726 | 70 | ||
AK395937.1 | - | 4 - 5 | 748 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
