Assay Details
Gene Symbol: C7H6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013977870.2 | XP_013833324.2 | 142 | 82 | ||
XM_021098650.1 | XP_020954309.1 | 594 | 82 | |||
GenBank mRNA | AK239396.1 | - | 1 - 2 | 79 | 82 | |
AK390756.1 | - | 1 - 2 | 87 | 82 | ||
AK390877.1 | - | 1 - 2 | 53 | 82 | ||
AK396826.1 | - | 1 - 2 | 82 | 82 | ||
AK398191.1 | - | 1 - 2 | 49 | 82 | ||
AK400725.1 | - | 1 - 2 | 110 | 82 | ||
AK400837.1 | - | 1 - 2 | 75 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013977870.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
