Assay Details
Gene Symbol: C7H6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013977868.2 | XP_013833322.2 | 994 | 106 | ||
XM_013977869.2 | XP_013833323.2 | 726 | 106 | |||
XM_013977870.2 | XP_013833324.2 | 659 | 106 | |||
XM_021098650.1 | XP_020954309.1 | 1170 | 106 | |||
XM_021098651.1 | XP_020954310.1 | 713 | 106 | |||
XM_021098652.1 | XP_020954311.1 | 1264 | 106 | |||
XM_021098653.1 | XP_020954312.1 | 585 | 106 | |||
GenBank mRNA | AK239396.1 | - | 4 - 5 | 655 | 106 | |
AK390756.1 | - | 4 - 5 | 663 | 106 | ||
AK390877.1 | - | 4 - 5 | 629 | 106 | ||
AK394782.1 | - | 5 - 6 | 780 | 106 | ||
AK398191.1 | - | 4 - 5 | 625 | 106 | ||
AK400725.1 | - | 4 - 5 | 627 | 106 | ||
AK400837.1 | - | 4 - 5 | 651 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
