Assay Details
Gene Symbol: C7H6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013977868.2 | XP_013833322.2 | 1131 | 89 | ||
XM_013977869.2 | XP_013833323.2 | 863 | 89 | |||
XM_013977870.2 | XP_013833324.2 | 796 | 89 | |||
XM_021098650.1 | XP_020954309.1 | 1307 | 89 | |||
XM_021098651.1 | XP_020954310.1 | 850 | 89 | |||
XM_021098652.1 | XP_020954311.1 | 1401 | 89 | |||
XM_021098653.1 | XP_020954312.1 | 722 | 89 | |||
GenBank mRNA | AK239396.1 | - | 5 - 6 | 792 | 89 | |
AK390756.1 | - | 5 - 6 | 800 | 89 | ||
AK390877.1 | - | 5 - 6 | 766 | 89 | ||
AK394782.1 | - | 6 - 7 | 917 | 89 | ||
AK396826.1 | - | 5 - 6 | 736 | 89 | ||
AK398191.1 | - | 5 - 6 | 762 | 89 | ||
AK400725.1 | - | 5 - 6 | 764 | 89 | ||
AK400837.1 | - | 5 - 6 | 788 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
