Assay Details
Gene Symbol: EIF2S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001928339.4 | XP_001928374.1 | 728 | 80 | ||
GenBank mRNA | AK239598.1 | - | 3 - 4 | 447 | 80 | |
AK351968.1 | - | 3 - 4 | 434 | 80 | ||
AK391511.1 | - | 3 - 4 | 80 | |||
AK394680.1 | - | 3 - 4 | 449 | 80 | ||
AY609835.1 | - | 3 - 4 | 420 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
