Assay Details
Gene Symbol: CD99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK239858.1 | - | 1 - 2 | 47 | 70 | |
AK352097.1 | - | 1 - 2 | 47 | 70 | ||
AK392781.1 | - | 3 - 4 | 214 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
