Assay Details
Gene Symbol: TRIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021088927.1 | XP_020944586.1 | 1317 | 97 | ||
GenBank mRNA | AK240605.1 | - | 1 - 2 | 857 | 97 | |
JX092266.1 | - | 1 - 2 | 78 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
