Assay Details
Gene Symbol: TMEM139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003134572.4 | XP_003134620.1 | 866 | 101 | ||
GenBank mRNA | AK343247.1 | - | 1 - 2 | 62 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003134572.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
