Assay Details
| Gene Symbol: DLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077714.1 | XP_020933373.1 | 4619 | 63 | ||
| XM_021077715.1 | XP_020933374.1 | 4784 | 63 | |||
| XM_021077716.1 | XP_020933375.1 | 4520 | 63 | |||
| XM_021077717.1 | XP_020933376.1 | 4457 | 63 | |||
| XM_021077718.1 | XP_020933377.1 | 4459 | 63 | |||
| XM_021077719.1 | XP_020933378.1 | 4325 | 63 | |||
| XM_021077720.1 | XP_020933379.1 | 4509 | 63 | |||
| XM_021077721.1 | XP_020933380.1 | 4675 | 63 | |||
| XM_021077722.1 | XP_020933381.1 | 3324 | 63 | |||
| XM_021077724.1 | XP_020933383.1 | 3081 | 63 | |||
| XM_021077725.1 | XP_020933384.1 | 3147 | 63 | |||
| XM_021077726.1 | XP_020933385.1 | 4657 | 63 | |||
| XM_021077727.1 | XP_020933386.1 | 10374 | 63 | |||
| XM_021077728.1 | XP_020933387.1 | 3830 | 63 | |||
| GenBank mRNA | AK343417.1 | - | 7 - 8 | 1695 | 63 | |
| AK345458.1 | - | 3 - 4 | 459 | 63 | ||
| AK389905.1 | - | 11 - 12 | 3033 | 63 | ||
| AK400197.1 | - | 11 - 12 | 2941 | 63 | ||
More Information
Set Membership: |
|

DLC1
Genomic Map