Assay Details
| Gene Symbol: NDUFB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135056.4 | XP_003135104.1 | 635 | 61 | ||
| XM_021080407.1 | XP_020936066.1 | 234 | 61 | |||
| GenBank mRNA | AK343497.1 | - | 1 - 2 | 217 | 61 | |
| AK348030.1 | - | 1 - 2 | 209 | 61 | ||
| AK399224.1 | - | 1 - 2 | 218 | 61 | ||
More Information
Set Membership: |
|

NDUFB11
Genomic Map