Assay Details
| Gene Symbol: ASAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134187.6 | XP_003134235.3 | 1160 | 73 | ||
| GenBank mRNA | AK343741.1 | - | 12 - 13 | 1118 | 73 | |
| AK389671.1 | - | 5 - 6 | 501 | 73 | ||
More Information
Set Membership: |
|

ASAH1
Genomic Map