Assay Details
Gene Symbol: ATP6V1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003134698.3 | XP_003134746.1 | 294 | 86 | ||
GenBank mRNA | AK343799.1 | - | 1 - 2 | 233 | 86 | |
AK343967.1 | - | 1 - 2 | 188 | 86 | ||
AK392017.1 | - | 1 - 2 | 219 | 86 | ||
AY610372.1 | - | 1 - 2 | 175 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
