Assay Details
Gene Symbol: TMEM267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021077012.1 | XP_020932671.1 | 524 | 78 | ||
XM_021077013.1 | XP_020932672.1 | 588 | 78 | |||
XM_021077014.1 | XP_020932673.1 | 553 | 78 | |||
XM_021077015.1 | XP_020932674.1 | 669 | 78 | |||
XM_021077016.1 | XP_020932675.1 | 571 | 78 | |||
XM_021077017.1 | XP_020932676.1 | 521 | 78 | |||
XM_021077018.1 | XP_020932677.1 | 529 | 78 | |||
XM_021077019.1 | XP_020932678.1 | 1197 | 78 | |||
XM_021077021.1 | XP_020932680.1 | 530 | 78 | |||
XM_021077022.1 | XP_020932681.1 | 1117 | 78 | |||
XM_021077023.1 | XP_020932682.1 | 523 | 78 | |||
XM_021077024.1 | XP_020932683.1 | 465 | 78 | |||
XM_021077025.1 | XP_020932684.1 | 418 | 78 | |||
GenBank mRNA | AK343831.1 | - | 2 - 3 | 422 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
