Assay Details
Gene Symbol: FILIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021092321.1 | XP_020947980.1 | 787 | 77 | ||
XM_021092328.1 | XP_020947987.1 | 787 | 77 | |||
XM_021092336.1 | XP_020947995.1 | 283 | 77 | |||
XM_021092343.1 | XP_020948002.1 | 787 | 77 | |||
GenBank mRNA | AK344037.1 | - | 2 - 3 | 514 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
